Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTBK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032538.1 | NP_115927.1 | 4 | 581 | ||
XM_011514947.2 | XP_011513249.1 | 3 | 371 | |||
XM_017011364.1 | XP_016866853.1 | 3 | 371 | |||
XM_017011365.1 | XP_016866854.1 | 3 | 371 | |||
XM_017011366.1 | XP_016866855.1 | 3 | 371 | |||
XM_017011367.1 | XP_016866856.1 | 3 | 371 | |||
GenBank mRNA | AB058758.1 | - | 2 | 150 | ||
AB218664.1 | - | 5 | 385 | |||
AK131217.1 | - | 3 | 311 |