Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLITRK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144003.2 | NP_001137475.1 | 5 | 742 | ||
NM_001144004.2 | NP_001137476.1 | 5 | 730 | |||
NM_001144005.2 | NP_001137477.1 | 5 | 752 | |||
NM_001144006.2 | NP_001137478.1 | 3 | 962 | |||
NM_001144008.2 | NP_001137480.1 | 3 | 974 | |||
NM_001144009.2 | NP_001137481.1 | 2 | 815 | |||
NM_001144010.2 | NP_001137482.1 | 3 | 952 | |||
NM_032539.4 | NP_115928.1 | 5 | 1225 | |||
XM_005262342.3 | XP_005262399.1 | 5 | 1827 | |||
XM_005262343.3 | XP_005262400.1 | 5 | 1837 | |||
XM_005262344.3 | XP_005262401.1 | 5 | 1815 | |||
XM_005262345.3 | XP_005262402.1 | 4 | 1678 | |||
GenBank mRNA | AB058757.1 | - | 1 | 4490 | ||
AK091015.1 | - | 1 | 343 | |||
AK096278.1 | - | 5 | 1221 | |||
AK289732.1 | - | 5 | 592 | |||
BC113011.2 | - | 3 | 816 | 40028691 | ||
BC113012.2 | - | 3 | 818 | 40028693 | ||
CR627412.1 | - | 2 | 2264 | |||
DA184412.1 | - | 3 | 522 | |||
DA332123.1 | - | 3 | 491 |