Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290325.1 | NP_001277254.1 | 9 | 2572 | ||
NM_001290326.1 | NP_001277255.1 | 10 | 2688 | |||
NM_032557.6 | NP_115946.2 | 9 | 2572 | |||
XM_011532360.2 | XP_011530662.1 | 8 | 1958 | |||
GenBank mRNA | AB067478.1 | - | 8 | 1253 | ||
AF211481.1 | - | 1 | 145 | |||
AK057992.1 | - | 6 | 1096 | |||
AK091712.1 | - | 1 | 442 | |||
AK093754.1 | - | 10 | 1911 | |||
AK126943.1 | - | 10 | 2400 | |||
AL833976.1 | - | 9 | 2544 | |||
BC039115.1 | - | 9 | 2572 | 5258804 | ||
BC068975.1 | - | 9 | 2519 | 6067501 |