Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032558.2 | NP_115947.2 | 11 | 1417 | ||
XM_006717305.3 | XP_006717368.1 | 10 | 1294 | |||
XM_011519114.2 | XP_011517416.1 | 11 | 1108 | |||
XM_017015217.1 | XP_016870706.1 | 11 | 1218 | |||
XM_017015218.1 | XP_016870707.1 | 11 | 1145 | |||
XM_017015219.1 | XP_016870708.1 | 10 | 955 | |||
XM_017015220.1 | XP_016870709.1 | 10 | 969 | |||
XM_017015221.1 | XP_016870710.1 | 10 | 1010 | |||
GenBank mRNA | AF255650.1 | - | 5 | 390 | ||
AK027659.1 | - | 7 | 722 | |||
AK222836.1 | - | 7 | 732 | |||
AK300615.1 | - | 9 | 891 | |||
AL832415.1 | - | 4 | 287 | |||
BC021093.1 | - | 6 | 527 | |||
BC106062.1 | - | 11 | 1154 | 6162855 | ||
BX648846.1 | - | 8 | 831 |