Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C22orf23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207062.1 | NP_001193991.1 | 4 | 408 | ||
NM_032561.4 | NP_115950.3 | 5 | 619 | |||
XM_005261781.1 | XP_005261838.1 | 5 | 583 | |||
XM_005261782.3 | XP_005261839.1 | 5 | 463 | |||
XM_005261783.2 | XP_005261840.1 | 4 | 520 | |||
XM_005261784.2 | XP_005261841.1 | 4 | 520 | |||
GenBank mRNA | AF324466.1 | - | 4 | 419 | ||
AK057349.1 | - | 5 | 669 | |||
BC031998.1 | - | 5 | 501 | 4755328 | ||
BI827388.1 | - | 4 | 408 | |||
BP368870.1 | - | 4 | 502 | |||
CR456440.1 | - | 5 | 499 |