Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032564.4 | NP_115953.2 | 2 | 465 | ||
XM_011545304.2 | XP_011543606.1 | 2 | 169 | |||
GenBank mRNA | AB048286.2 | - | 2 | 436 | ||
AB073384.1 | - | 1 | 13 | |||
AF384161.2 | - | 2 | 213 | |||
AL834287.1 | - | 2 | 344 | |||
AY358532.1 | - | 2 | 390 | |||
BC015234.1 | - | 2 | 454 | 3903313 |