Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032567.3 | NP_115956.3 | 1 | 345 | ||
GenBank mRNA | AF333098.1 | - | 1 | 316 | ||
AK098575.1 | - | 1 | 342 | |||
AK314007.1 | - | 1 | 344 | |||
BC033798.1 | - | 1 | 292 | 5172551 | ||
BG723698.1 | - | 1 | 353 | |||
BU853967.1 | - | 1 | 315 |