Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289158.1 | NP_001276087.1 | 14 | 1835 | ||
NM_001289159.1 | NP_001276088.1 | 12 | 1613 | |||
NM_032571.4 | NP_115960.2 | 15 | 1991 | |||
XM_011528374.2 | XP_011526676.1 | 14 | 1882 | |||
XM_017027383.1 | XP_016882872.1 | 10 | 1285 | |||
GenBank mRNA | AF239764.1 | - | 15 | 1928 | ||
AK290909.1 | - | 15 | 1915 | |||
AK298653.1 | - | 12 | 1539 | |||
AL832663.1 | - | 15 | 1992 | |||
AY358817.1 | - | 14 | 1732 |
Set Membership: |
G-protein coupled receptor |