Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256267.1 | NP_001243196.1 | 14 | 2944 | ||
NM_001256268.1 | NP_001243197.1 | 17 | 3353 | |||
NM_032578.3 | NP_115967.2 | 13 | 3049 | |||
NR_045662.3 | 13 | 2184 | ||||
NR_045663.3 | 11 | 2886 | ||||
XM_017016833.1 | XP_016872322.1 | 14 | 3385 | |||
XM_017016834.1 | XP_016872323.1 | 13 | 2843 | |||
GenBank mRNA | AF328296.1 | - | 13 | 3244 | ||
AL832002.2 | - | 13 | 2972 | |||
AL832379.1 | - | 13 | 2146 | |||
AL834247.2 | - | 13 | 2971 | |||
BC144334.1 | - | 11 | 2648 | |||
BX647637.1 | - | 13 | 2980 | |||
BX647640.1 | - | 14 | 2937 |