Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013399.1 | NP_001013417.1 | 11 | 799 | ||
NM_005190.3 | NP_005181.2 | 11 | 1021 | |||
XM_006715594.1 | XP_006715657.1 | 11 | 1180 | |||
GenBank mRNA | AB125137.1 | - | 11 | 808 | ||
AK222768.1 | - | 11 | 832 | |||
AK223247.1 | - | 11 | 822 | |||
AK291914.1 | - | 11 | 830 | |||
AK298558.1 | - | 10 | 760 | |||
BC010135.2 | - | 11 | 854 | 4106819 | ||
BC041123.1 | - | 11 | 1004 | 5759974 | ||
BC056153.1 | - | 11 | 843 | 6157106 | ||
M74091.1 | - | 11 | 822 |