Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032588.3 | NP_115977.2 | 3, 4 | 649 | ||
XM_017002559.1 | XP_016858048.1 | 3, 4 | 649 | |||
GenBank mRNA | AF353673.1 | - | 3, 4 | 628 | ||
AJ276484.1 | - | 3, 4 | 632 | |||
AK056942.1 | - | 3, 4 | 645 | |||
AK297820.1 | - | 3, 4 | 561 | |||
BC080529.1 | - | 3, 4 | 627 | 4862321 | ||
BT007373.1 | - | 3, 4 | 509 |