Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005366.1 | NP_001005366.1 | 10 | 1475 | ||
NM_032590.4 | NP_115979.3 | 10 | 1265 | |||
XM_005253955.3 | XP_005254012.1 | 10 | 1475 | |||
XM_005253956.3 | XP_005254013.1 | 10 | 1072 | |||
XM_011538867.2 | XP_011537169.1 | 10 | 2518 | |||
XM_011538868.2 | XP_011537170.1 | 10 | 2514 | |||
XM_011538869.2 | XP_011537171.1 | 10 | 2516 | |||
GenBank mRNA | AJ459424.1 | - | 10 | 1237 | ||
AK127328.1 | - | 10 | 1475 | |||
AL133572.1 | - | 6 | 778 | |||
BC115379.1 | - | 9 | 1125 | |||
BC115380.1 | - | 10 | 1204 | 40029132 | ||
BX648928.1 | - | 8 | 997 |