Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127219.1 | NP_001120691.1 | 10 | 1054 | ||
NM_032592.3 | NP_115981.1 | 10 | 1350 | |||
XM_005253170.3 | XP_005253227.2 | 10 | 1493 | |||
XM_005253172.2 | XP_005253229.1 | 7 | 581 | |||
XM_006718348.2 | XP_006718411.1 | 10 | 802 | |||
XM_011520406.2 | XP_011518708.1 | 10 | 1496 | |||
XM_011520407.1 | XP_011518709.1 | 10 | 846 | |||
XM_011520410.2 | XP_011518712.1 | 10 | 1747 | |||
GenBank mRNA | AK057649.1 | - | 10 | 1344 | ||
AL833533.1 | - | 2 | 2347 | |||
AY026508.1 | - | 10 | 1305 | |||
BC020197.1 | - | 10 | 1324 | 3632065 | ||
KU178811.1 | - | 9 | 910 | |||
KU178812.1 | - | 9 | 910 | |||
KU178813.1 | - | 8 | 850 |