Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INSM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032594.3 | NP_115983.3 | 1 | 1649 | ||
GenBank mRNA | AB037912.1 | - | 1 | 1524 | ||
AF260323.1 | - | 1 | 1652 | |||
AK096639.1 | - | 1 | 1524 | |||
BC130377.1 | - | 1 | 1530 | 40146408 | ||
BC130379.1 | - | 1 | 1530 | 40146410 | ||
JX403968.1 | - | 1 | 1438 |