Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252195.1 | NP_001239124.1 | 4 | 451 | ||
NM_032596.3 | NP_115985.2 | 6 | 985 | |||
NM_147168.1 | NP_671697.1 | 3 | 527 | |||
NM_147169.2 | NP_671698.1 | 3 | 373 | |||
XM_005251616.4 | XP_005251673.1 | 6 | 3972 | |||
XM_005251618.4 | XP_005251675.1 | 4 | 519 | |||
XM_011518055.2 | XP_011516357.1 | 4 | 793 | |||
GenBank mRNA | AF367474.1 | - | 3 | 312 | ||
AK058087.1 | - | 6 | 923 | |||
AK094751.1 | - | 2 | 1685 | |||
AK225981.1 | - | 3 | 445 | |||
AW961020.1 | - | 3 | 527 | |||
BC022084.1 | - | 6 | 970 | 4825995 | ||
BC029484.1 | - | 6 | 985 | 5271301 | ||
BI561111.1 | - | 4 | 416 | |||
CB854584.1 | - | 3 | 496 |