Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079692.2 | NP_001073160.1 | 1 | 615 | ||
NM_001261827.1 | NP_001248756.1 | 1 | 615 | |||
NM_001261828.1 | NP_001248757.1 | 1 | 615 | |||
NM_032597.4 | NP_115986.3 | 1 | 615 | |||
NR_049731.1 | 1 | 615 | ||||
NR_049732.1 | 1 | 615 | ||||
NR_049733.1 | 1 | 615 | ||||
NR_049734.1 | 1 | 615 | ||||
GenBank mRNA | AF367473.2 | - | 1 | 126 | ||
AK314674.1 | - | 1 | 327 | |||
AY094610.1 | - | 1 | 199 | |||
AY094611.1 | - | 1 | 199 | |||
AY584610.1 | - | 1 | 199 | |||
BC107431.1 | - | 1 | 122 | |||
DB082055.1 | - | 1 | 165 | |||
DB450359.2 | - | 1 | 123 | |||
DB727789.1 | - | 1 | 133 |