Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM71F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282788.1 | NP_001269717.1 | 3 | 619 | ||
NM_001282789.1 | NP_001269718.1 | 4 | 566 | |||
NM_032599.3 | NP_115988.1 | 3 | 619 | |||
NR_104242.1 | 3 | 619 | ||||
NR_104243.1 | 4 | 508 | ||||
XM_017012743.1 | XP_016868232.1 | 3 | 662 | |||
GenBank mRNA | AF367470.1 | - | 3 | 572 | ||
AK093161.1 | - | 4 | 563 | |||
AK310308.1 | - | 3 | 566 | |||
AK310426.1 | - | 4 | 475 | |||
AK313608.1 | - | 3 | 570 | |||
BC037248.1 | - | 3 | 558 | 5165912 |