Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM71F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282788.1 | NP_001269717.1 | 6 | 1007 | ||
NM_001282789.1 | NP_001269718.1 | 7 | 954 | |||
NM_032599.3 | NP_115988.1 | 6 | 1013 | |||
NR_104242.1 | 7 | 1311 | ||||
NR_104243.1 | 8 | 1200 | ||||
GenBank mRNA | AF367470.1 | - | 6 | 966 | ||
AK093161.1 | - | 7 | 951 | |||
AK313608.1 | - | 6 | 958 | |||
BC037248.1 | - | 6 | 946 | 5165912 |