Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAXX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141969.1 | NP_001135441.1 | 6 | 1804 | ||
NM_001141970.1 | NP_001135442.1 | 6 | 1735 | |||
NM_001254717.1 | NP_001241646.1 | 5 | 1545 | |||
NM_001350.4 | NP_001341.1 | 6 | 1801 | |||
XM_005248860.3 | XP_005248917.1 | 4 | 651 | |||
GenBank mRNA | AB209493.1 | - | 2 | 2269 | ||
AF006041.1 | - | 1 | 122 | |||
AF015956.2 | - | 5 | 1651 | |||
AF039136.1 | - | 6 | 1744 | |||
AF050179.1 | - | 6 | 1712 | |||
AF097742.1 | - | 5 | 1604 | |||
AK223404.1 | - | 6 | 1802 | |||
AK292187.1 | - | 6 | 1802 | |||
AK303767.1 | - | 6 | 1678 | |||
AK303854.1 | - | 5 | 1502 | |||
BC000220.1 | - | 599 | ||||
BC109073.1 | - | 6 | 1687 | 40034214 | ||
BC109074.1 | - | 6 | 1687 | 40034219 | ||
CR457085.1 | - | 5 | 1597 | |||
HQ436528.1 | - | 5 | 1597 | |||
HQ436529.1 | - | 5 | 1597 |