Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168399.1 | NP_001161871.1 | 3 | 574 | ||
NM_001168400.1 | NP_001161872.1 | 3 | 571 | |||
NM_001168401.1 | NP_001161873.1 | 3 | 316 | |||
NM_032621.3 | NP_116010.1 | 3 | 388 | |||
GenBank mRNA | AF251053.1 | - | 3 | 220 | ||
AK312085.1 | - | 3 | 312 | |||
AY833560.1 | - | 3 | 320 | |||
BC015522.1 | - | 3 | 266 | 3914401 | ||
BI670202.1 | - | 3 | 367 | |||
BU569325.1 | - | 3 | 391 | |||
BU596902.1 | - | 3 | 299 | |||
CB961647.1 | - | 3 | 580 | |||
CK001963.1 | - | 3 | 319 |