Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDGFRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001520.2 | NP_001001520.1 | 12 | 1605 | ||
NM_032631.3 | NP_116020.1 | 12 | 1605 | |||
XM_011528375.2 | XP_011526677.1 | 12 | 1965 | |||
XM_011528376.2 | XP_011526678.1 | 12 | 1962 | |||
XM_011528377.2 | XP_011526679.1 | 12 | 1965 | |||
GenBank mRNA | AB208892.1 | - | 10 | 2343 | ||
AF294267.1 | - | 12 | 1512 | |||
AY358600.1 | - | 12 | 1514 | |||
BC000755.2 | - | 12 | 1495 | 3507276 | ||
BC009449.1 | - | 9 | 1144 |