Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032639.3 | NP_116028.1 | 13 | 2106 | ||
XM_011515592.2 | XP_011513894.1 | 14 | 2116 | |||
GenBank mRNA | BC002838.2 | - | 13 | 1795 | 3635744 |