Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301175.1 | NP_001288104.1 | 7 | 894 | ||
NM_032663.4 | NP_116052.2 | 4 | 520 | |||
XM_005253962.3 | XP_005254019.1 | 4 | 506 | |||
XM_005253965.4 | XP_005254022.1 | 3 | 331 | |||
XM_006719653.3 | XP_006719716.1 | 5 | 593 | |||
XM_011538894.2 | XP_011537196.1 | 2 | 227 | |||
XM_017020048.1 | XP_016875537.1 | 4 | 515 | |||
XM_017020049.1 | XP_016875538.1 | 4 | 506 | |||
XM_017020050.1 | XP_016875539.1 | 4 | 615 | |||
XM_017020051.1 | XP_016875540.1 | 5 | 471 | |||
XM_017020052.1 | XP_016875541.1 | 4 | 517 | |||
XM_017020053.1 | XP_016875542.1 | 3 | 247 | |||
XM_017020054.1 | XP_016875543.1 | 2 | 125 | |||
GenBank mRNA | AJ586136.1 | - | 4 | 411 | ||
AK027820.1 | - | 4 | 485 | |||
AK097830.1 | - | 7 | 894 | |||
BC004868.2 | - | 4 | 510 | 3833242 | ||
BC022094.2 | - | 2 | 62 | |||
HY127211.1 | - | 2 | 228 |