Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301175.1 | NP_001288104.1 | 10, 11 | 1208 | ||
NM_032663.4 | NP_116052.2 | 7, 8 | 834 | |||
XM_005253962.3 | XP_005254019.1 | 7, 8 | 817 | |||
XM_005253965.4 | XP_005254022.1 | 6, 7 | 645 | |||
XM_006719653.3 | XP_006719716.1 | 8, 9 | 907 | |||
XM_011538894.2 | XP_011537196.1 | 5, 6 | 538 | |||
XM_017020048.1 | XP_016875537.1 | 7, 8 | 829 | |||
XM_017020049.1 | XP_016875538.1 | 7, 8 | 817 | |||
XM_017020050.1 | XP_016875539.1 | 7, 8 | 929 | |||
XM_017020051.1 | XP_016875540.1 | 8, 9 | 785 | |||
XM_017020052.1 | XP_016875541.1 | 7, 8 | 831 | |||
XM_017020053.1 | XP_016875542.1 | 6, 7 | 561 | |||
XM_017020054.1 | XP_016875543.1 | 5, 6 | 436 | |||
GenBank mRNA | AJ586136.1 | - | 7, 8 | 725 | ||
AK027820.1 | - | 7, 8 | 799 | |||
AK097830.1 | - | 10, 11 | 1208 | |||
BC004868.2 | - | 7, 8 | 824 | 3833242 | ||
BC022094.2 | - | 5, 6 | 376 | |||
CN367077.1 | - | 1, 2 | 98 |