Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBA1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303114.1 | NP_001290043.1 | 4 | 1660 | ||
NM_001303115.1 | NP_001290044.1 | 6 | 1893 | |||
NM_001303116.1 | NP_001290045.1 | 5 | 1775 | |||
NM_001303117.1 | NP_001290046.1 | 5 | 1778 | |||
NM_032704.4 | NP_116093.1 | 4 | 1605 | |||
GenBank mRNA | AK223024.1 | - | 4 | 1522 | ||
AK293589.1 | - | 4 | 1660 | |||
BC004949.1 | - | 4 | 1502 | 3617714 | ||
BC005946.1 | - | 4 | 1522 | 4134187 | ||
BC011790.1 | - | 4 | 1503 | 4025017 | ||
BC019298.2 | - | 4 | 1484 | 2905506 | ||
BC021088.1 | - | 4 | 1483 | 3543481 | ||
BC033064.1 | - | 5 | 1020 | 3926854 | ||
BC051297.1 | - | 4 | 1498 | 3832363 | ||
BC063036.1 | - | 4 | 1544 | 4397636 |