Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYROXD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032709.2 | NP_116098.2 | 8 | 796 | ||
XM_011540293.2 | XP_011538595.1 | 9 | 2218 | |||
XM_017016835.1 | XP_016872324.1 | 8 | 757 | |||
XM_017016836.1 | XP_016872325.1 | 8 | 757 | |||
XM_017016837.1 | XP_016872326.1 | 8 | 757 | |||
XM_017016838.1 | XP_016872327.1 | 8 | 2992 | |||
XM_017016839.1 | XP_016872328.1 | 6 | 849 | |||
XM_017016840.1 | XP_016872329.1 | 6 | 703 | |||
XM_017016841.1 | XP_016872330.1 | 5 | 1010 | |||
GenBank mRNA | AK074429.1 | - | 8 | 757 | ||
AK075265.1 | - | 8 | 734 | |||
AK225637.1 | - | 8 | 750 | |||
AK295699.1 | - | 8 | 769 | |||
BC006131.1 | - | 8 | 759 | 3626506 |
Gene Symbol: MIR1287 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_031619.1 | 1 | 37 |