Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032726.3 | NP_116115.1 | 11 | 1887 | ||
XM_005246913.2 | XP_005246970.1 | 11 | 1849 | |||
XM_011512012.1 | XP_011510314.1 | 12 | 1945 | |||
XM_017005114.1 | XP_016860603.1 | 12 | 1886 | |||
XM_017005115.1 | XP_016860604.1 | 12 | 1945 | |||
XM_017005116.1 | XP_016860605.1 | 12 | 1945 | |||
XM_017005117.1 | XP_016860606.1 | 11 | 1792 | |||
XM_017005118.1 | XP_016860607.1 | 12 | 2772 | |||
XM_017005119.1 | XP_016860608.1 | 9 | 1243 | |||
XM_017005120.1 | XP_016860609.1 | 5 | 655 | |||
GenBank mRNA | AB209599.1 | - | 5 | 2581 | ||
AK023083.1 | - | 10 | 1583 | |||
AK223203.1 | - | 11 | 1753 | |||
AK297808.1 | - | 6 | 683 | |||
AK299458.1 | - | 11 | 1754 | |||
AY512961.1 | - | 8 | 1297 | |||
BC006355.2 | - | 11 | 1743 | 4124286 |