Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032731.3 | NP_116120.1 | 3 | 467 | ||
GenBank mRNA | AF086210.1 | - | 3 | 318 | ||
AJ344101.1 | - | 3 | 362 | |||
AK291963.1 | - | 3 | 351 | |||
AK311853.1 | - | 3 | 322 | |||
BC006405.2 | - | 3 | 345 | 4298953 |