Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032737.3 | NP_116126.3 | 8, 9 | 1510 | ||
GenBank mRNA | BC006551.2 | - | 8, 9 | 1475 | 2961134 | |
BT007441.1 | - | 8, 9 | 1415 | |||
KP137589.1 | - | 5, 6 | 1064 | |||
M94362.1 | - | 7, 8 | 1160 |