Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079862.2 | NP_001073331.1 | 4 | 531 | ||
NM_001079863.1 | NP_001073332.1 | 4 | 651 | |||
NM_001178017.1 | NP_001171488.1 | 4 | 670 | |||
NM_001178041.2 | NP_001171512.1 | 5 | 657 | |||
NM_001178042.2 | NP_001171513.1 | 5 | 612 | |||
NM_001178043.2 | NP_001171514.1 | 4 | 561 | |||
NM_001282633.1 | NP_001269562.1 | 5 | 647 | |||
NM_001282634.1 | NP_001269563.1 | 5 | 617 | |||
NM_001282635.1 | NP_001269564.1 | 5 | 647 | |||
NM_001282636.1 | NP_001269565.1 | 4 | 561 | |||
NM_020548.7 | NP_065438.1 | 5 | 642 | |||
NR_104221.1 | 4 | 810 | ||||
GenBank mRNA | BC006466.1 | - | 4 | 446 | ||
BC062996.1 | - | 5 | 611 | 5168729 | ||
BM768076.1 | - | 4 | 478 | |||
BP294452.1 | - | 4 | 473 | |||
BP318721.1 | - | 3 | 335 | |||
M14200.1 | - | 4 | 473 |