Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USMG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206426.1 | NP_001193355.1 | 2 | 190 | ||
NM_001206427.1 | NP_001193356.1 | 3 | 477 | |||
NM_032747.3 | NP_116136.1 | 3 | 386 | |||
GenBank mRNA | AJ272056.1 | - | 2 | 160 | ||
AK311888.1 | - | 2 | 130 | |||
AL110185.1 | - | 2 | 129 | |||
AY740522.1 | - | 1 | 76 | |||
BC007087.1 | - | 3 | 339 | 4245636 | ||
BC072683.1 | - | 2 | 137 | 6056764 | ||
BC091507.1 | - | 2 | 135 | 30417704 | ||
CN367000.1 | - | 3 | 423 | |||
DW446355.1 | - | 2 | 206 |