Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319074.1 | NP_001306003.1 | 3 | 1766 | ||
NM_032753.3 | NP_116142.1 | 3 | 1507 | |||
GenBank mRNA | AY211277.1 | - | 3 | 1499 | ||
BC007284.1 | - | 4 | 1404 | 3344166 | ||
BC018709.1 | - | 3 | 1766 | 4635722 | ||
BC032512.1 | - | 3 | 1777 | 5491469 |