Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004395.3 | NP_004386.2 | 6 | 662 | ||
NM_080881.2 | NP_543157.1 | 7 | 778 | |||
XM_005265827.3 | XP_005265884.1 | 6 | 609 | |||
XM_006714826.2 | XP_006714889.1 | 6 | 499 | |||
XM_011534446.2 | XP_011532748.1 | 7 | 781 | |||
XM_011534447.2 | XP_011532749.1 | 6 | 1278 | |||
XM_017009139.1 | XP_016864628.1 | 6 | 780 | |||
XM_017009140.1 | XP_016864629.1 | 6 | 771 | |||
GenBank mRNA | AK094125.1 | - | 5 | 708 | ||
AK225523.1 | - | 6 | 603 | |||
AK314645.1 | - | 6 | 603 | |||
AL110225.1 | - | 7 | 1107 | |||
BC000283.1 | - | 6 | 586 | 3356428 | ||
BC007281.1 | - | 6 | 586 | 3343626 | ||
BC007567.1 | - | 6 | 586 | 3028196 | ||
D17530.1 | - | 6 | 587 | |||
U00802.1 | - | 6 | 562 |