Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032775.3 | NP_116164.2 | 4 | 614 | ||
NR_033825.1 | 3 | 448 | ||||
XM_017029018.1 | XP_016884507.1 | 3 | 782 | |||
XM_017029019.1 | XP_016884508.1 | 4 | 910 | |||
XM_017029020.1 | XP_016884509.1 | 5 | 2089 | |||
XM_017029021.1 | XP_016884510.1 | 4 | 543 | |||
XM_017029022.1 | XP_016884511.1 | 3 | 714 | |||
XM_017029023.1 | XP_016884512.1 | 3 | 3174 | |||
XM_017029024.1 | XP_016884513.1 | 3 | 746 | |||
XM_017029025.1 | XP_016884514.1 | 3 | 262 | |||
GenBank mRNA | AK027266.1 | - | 4 | 553 | ||
AK222850.1 | - | 4 | 520 | |||
AK290669.1 | - | 4 | 526 | |||
AK294682.1 | - | 3 | 360 | |||
BC015923.1 | - | 4 | 566 | 4562420 | ||
CR456352.1 | - | 3 | 491 |