Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032783.4 | NP_116172.2 | 3 | 595 | ||
XM_005263315.2 | XP_005263372.1 | 3 | 562 | |||
XM_005263316.3 | XP_005263373.1 | 3 | 562 | |||
XM_006714391.1 | XP_006714454.1 | 3 | 562 | |||
XM_006714392.3 | XP_006714455.1 | 3 | 562 | |||
XM_011532385.1 | XP_011530687.1 | 3 | 562 | |||
XM_011532386.2 | XP_011530688.1 | 2 | 212 | |||
XM_017008782.1 | XP_016864271.1 | 3 | 562 | |||
XM_017008783.1 | XP_016864272.1 | 3 | 562 | |||
GenBank mRNA | AK027337.1 | - | 3 | 532 | ||
AK291756.1 | - | 3 | 532 | |||
AL833393.1 | - | 3 | 535 | |||
AV704400.1 | - | 3 | 561 | |||
BC021973.2 | - | 3 | 514 | 3625459 | ||
BC033650.1 | - | 3 | 525 | 5574637 | ||
BQ230058.1 | - | 3 | 549 |