Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316978.1 | NP_001303907.1 | 2 | 866 | ||
NM_001316979.1 | NP_001303908.1 | 2 | 593 | |||
NM_001316980.1 | NP_001303909.1 | 2 | 416 | |||
NM_001316981.1 | NP_001303910.1 | 2 | 718 | |||
NM_001316982.1 | NP_001303911.1 | 2 | 536 | |||
NM_032792.3 | NP_116181.1 | 2 | 622 | |||
XM_006723445.3 | XP_006723508.1 | 2 | 636 | |||
GenBank mRNA | AK027392.1 | - | 2 | 405 | ||
BC024738.1 | - | 2 | 617 | 5110354 | ||
DB043101.1 | - | 2 | 536 | |||
DR158681.1 | - | 2 | 492 | |||
HY023937.1 | - | 2 | 416 |