Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048164.2 | NP_001041629.1 | 6 | 1075 | ||
NM_032803.5 | NP_116192.4 | 6 | 1099 | |||
XM_017029912.1 | XP_016885401.1 | 5 | 1590 | |||
GenBank mRNA | AF320612.1 | - | 6 | 1050 | ||
AK027447.1 | - | 4 | 421 | |||
AK074865.1 | - | 6 | 1070 | |||
AK075014.1 | - | 6 | 1075 | |||
BC033816.1 | - | 6 | 1054 | 5206252 |