Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048164.2 | NP_001041629.1 | 12 | 1931 | ||
NM_032803.5 | NP_116192.4 | 12 | 1955 | |||
XM_017029912.1 | XP_016885401.1 | 11 | 2446 | |||
GenBank mRNA | AF320612.1 | - | 12 | 1906 | ||
AK074865.1 | - | 12 | 1926 | |||
AK075014.1 | - | 12 | 1931 | |||
BC033816.1 | - | 12 | 1910 | 5206252 |