Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032805.2 | NP_116194.2 | 2, 3 | 599 | ||
XM_017023791.1 | XP_016879280.1 | 1, 2 | 635 | |||
XM_017023792.1 | XP_016879281.1 | 1, 2 | 635 | |||
GenBank mRNA | AK027455.1 | - | 1, 2 | 327 | ||
CX781288.1 | - | 2, 3 | 599 |