Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005215.3 | NP_005206.2 | 22 | 3831 | ||
XM_011525843.1 | XP_011524145.1 | 22 | 3831 | |||
XM_011525844.2 | XP_011524146.1 | 19 | 2719 | |||
XM_017025568.1 | XP_016881057.1 | 22 | 3831 | |||
XM_017025569.1 | XP_016881058.1 | 22 | 3771 | |||
XM_017025570.1 | XP_016881059.1 | 18 | 2727 | |||
GenBank mRNA | AK310385.1 | - | 19 | 2385 | ||
X76132.1 | - | 22 | 3215 |