Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINGO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301186.1 | NP_001288115.1 | 6 | 1729 | ||
NM_001301187.1 | NP_001288116.1 | 6 | 1746 | |||
NM_001301189.1 | NP_001288118.1 | 6 | 1740 | |||
NM_001301191.1 | NP_001288120.1 | 6 | 1925 | |||
NM_001301192.1 | NP_001288121.1 | 5 | 1863 | |||
NM_001301194.1 | NP_001288123.1 | 4 | 1681 | |||
NM_001301195.1 | NP_001288124.1 | 4 | 1616 | |||
NM_001301197.1 | NP_001288126.1 | 4 | 1629 | |||
NM_001301198.1 | NP_001288127.1 | 3 | 1426 | |||
NM_001301199.1 | NP_001288128.1 | 4 | 1599 | |||
NM_001301200.1 | NP_001288129.1 | 4 | 1672 | |||
NM_032808.6 | NP_116197.4 | 2 | 1444 | |||
XM_011522118.2 | XP_011520420.1 | 2 | 1356 | |||
XM_017022682.1 | XP_016878171.1 | 7 | 2072 | |||
GenBank mRNA | AK027500.1 | - | 1 | 377 | ||
AK291363.1 | - | 2 | 1410 | |||
AL834260.1 | - | 1 | 1104 | |||
AY324320.1 | - | 2 | 1284 | |||
AY324322.1 | - | 1 | 780 | |||
AY324323.1 | - | 1 | 908 | |||
AY358284.1 | - | 2 | 1264 | |||
BC011057.1 | - | 2 | 1284 | 4214343 | ||
BC068558.1 | - | 4 | 1657 | 4797856 |