Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321967.1 | NP_001308896.1 | 7 | 810 | ||
NM_001321969.1 | NP_001308898.1 | 8 | 900 | |||
NM_032810.3 | NP_116199.2 | 7 | 1182 | |||
NR_135914.1 | 7 | 810 | ||||
XM_005270252.4 | XP_005270309.1 | 7 | 904 | |||
XM_011540302.1 | XP_011538604.1 | 5 | 575 | |||
XM_011540303.2 | XP_011538605.1 | 7 | 857 | |||
XM_017016847.1 | XP_016872336.1 | 7 | 1051 | |||
XM_017016849.1 | XP_016872338.1 | 8 | 1023 | |||
XM_017016850.1 | XP_016872339.1 | 8 | 1074 | |||
GenBank mRNA | AF361493.1 | - | 6 | 1016 | ||
AK027506.1 | - | 4 | 410 | |||
AK075223.1 | - | 7 | 1111 | |||
AL834370.1 | - | 6 | 595 | |||
BC010868.1 | - | 3 | 245 | |||
BC063530.1 | - | 6 | 641 | |||
BC073998.1 | - | 6 | 753 | 6199813 |