Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032811.2 | NP_116200.2 | 9 | 1405 | ||
NR_016021.1 | 8 | 1268 | ||||
XM_005271709.3 | XP_005271766.1 | 6 | 1211 | |||
XM_017018442.1 | XP_016873931.1 | 6 | 2226 | |||
GenBank mRNA | AK027527.1 | - | 8 | 1239 | ||
AK074140.1 | - | 8 | 992 | |||
AK222936.1 | - | 8 | 1244 | |||
AL713631.1 | - | 5 | 510 | |||
AL831881.1 | - | 3 | 245 | |||
AY696294.1 | - | 7 | 773 | |||
BC041043.1 | - | 8 | 1288 | |||
BC064392.1 | - | 8 | 1217 | |||
BC107428.1 | - | 8 | 1258 | |||
BC109269.2 | - | 7 | 887 | |||
DQ144542.1 | - | 9 | 1167 |