Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032811.2 | NP_116200.2 | 6 | 1054 | ||
NR_016021.1 | 5 | 917 | ||||
XM_005271709.3 | XP_005271766.1 | 3 | 860 | |||
XM_017018442.1 | XP_016873931.1 | 3 | 1875 | |||
GenBank mRNA | AK027527.1 | - | 5 | 888 | ||
AK074140.1 | - | 5 | 641 | |||
AK222936.1 | - | 5 | 893 | |||
AL713631.1 | - | 2 | 159 | |||
AY696294.1 | - | 4 | 422 | |||
BC041043.1 | - | 5 | 902 | |||
BC064392.1 | - | 5 | 866 | |||
BC107428.1 | - | 5 | 907 | |||
BC109269.2 | - | 4 | 536 | |||
DQ144542.1 | - | 6 | 816 |