Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNFT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109903.1 | NP_001103373.1 | 6, 7 | 966 | ||
NM_032814.3 | NP_116203.2 | 6, 7 | 966 | |||
GenBank mRNA | AK027533.1 | - | 7, 8 | 1002 | ||
AK093164.1 | - | 2, 3 | 767 | |||
BQ676121.1 | - | 3, 4 | 636 |