Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFATC2IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032815.3 | NP_116204.3 | 2 | 421 | ||
GenBank mRNA | AF458593.1 | - | 2 | 431 | ||
AK074761.1 | - | 2 | 411 | |||
AK304172.1 | - | 2 | 411 | |||
BC068007.1 | - | 2 | 47 | |||
BC080628.1 | - | 2 | 88 | |||
BC101741.1 | - | 2 | 481 | 8069247 | ||
BC112182.1 | - | 2 | 481 | 8327650 |