Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFATC2IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032815.3 | NP_116204.3 | 3 | 560 | ||
GenBank mRNA | AF458593.1 | - | 3 | 570 | ||
AK074761.1 | - | 3 | 550 | |||
BC018311.1 | - | 1 | 64 | |||
BC080628.1 | - | 3 | 227 | |||
BC101741.1 | - | 3 | 620 | 8069247 | ||
BC112182.1 | - | 3 | 620 | 8327650 |