Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM136A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032822.2 | NP_116211.2 | 3 | 437 | ||
XM_005264620.1 | XP_005264677.1 | 4 | 487 | |||
GenBank mRNA | AK027574.1 | - | 3 | 418 | ||
AK225148.1 | - | 3 | 374 | |||
AK225151.1 | - | 3 | 405 | |||
AK309543.1 | - | 3 | 1028 | |||
AK315877.1 | - | 4 | 601 | |||
AL833147.1 | - | 3 | 380 | |||
BC014975.1 | - | 3 | 395 | 4903550 | ||
DB266394.1 | - | 3 | 439 |