Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATOH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032827.6 | NP_116216.2 | 1 | 999 | ||
XM_006712122.3 | XP_006712185.1 | 1 | 999 | |||
XM_011533139.1 | XP_011531441.1 | 1 | 999 | |||
GenBank mRNA | AF529205.1 | - | 1 | 595 | ||
AK027614.1 | - | 1 | 792 | |||
AK074681.1 | - | 1 | 891 | |||
AL831857.1 | - | 1 | 138 | |||
BC021207.2 | - | 1 | 890 | 4109799 | ||
BC094832.1 | - | 1 | 669 | 6292270 |