Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318391.1 | NP_001305320.1 | 6 | 670 | ||
NM_032830.2 | NP_116219.2 | 6 | 724 | |||
XM_005256205.3 | XP_005256262.2 | 2 | 255 | |||
GenBank mRNA | AB075868.2 | - | 6 | 662 | ||
AK027419.1 | - | 3 | 279 | |||
AK027445.1 | - | 4 | 347 | |||
AK027634.1 | - | 6 | 676 | |||
AK027675.1 | - | 6 | 673 | |||
AK074795.1 | - | 6 | 690 | |||
AK310104.1 | - | 6 | 626 | |||
BC000167.2 | - | 2 | 169 | |||
BC009348.2 | - | 6 | 656 | 4131625 | ||
BX647265.1 | - | 6 | 638 |